Sterile Solution Bottles
About this item
Store solutions securely with CellPro™ Solution Bottles, individually wrapped and sterile to ensure optimal storage conditions for laboratory reagents.
- Compatible with CellPro™ Bottle Top Filtration Filters
- Lightweight yet durable wall construction
- Wide opening for quick access and non-drip pouring/decanting
- Non-pyrogenic
- Polystyrene (PS) bottles are packaged sterile, and individually wrapped
- Graduated in 25 ml molded increments for accurate filling
- Single packed sterile bottle and single packed sterile cap
- Eash case features lot number identification
CellPro™ Solution Bottles offer a sterile and convenient solution for various laboratory applications. Each bottle is individually wrapped to maintain sterility and ensure reliable performance. Designed for versatility, these solution bottles are suitable for storing and dispensing various solutions used in laboratory experiments, research, and clinical settings. With their sterile packaging and robust construction, CellPro™ Solution Bottles provide peace of mind and reliability for any laboratory's needs.
5 Options Available Below
Product Details & Documents
Store solutions securely with CellPro™ Solution Bottles, individually wrapped and sterile to ensure optimal storage conditions for laboratory reagents.
- Compatible with CellPro™ Bottle Top Filtration Filters
- Lightweight yet durable wall construction
- Wide opening for quick access and non-drip pouring/decanting
- Non-pyrogenic
- Polystyrene (PS) bottles are packaged sterile, and individually wrapped
- Graduated in 25 ml molded increments for accurate filling
- Single packed sterile bottle and single packed sterile cap
- Eash case features lot number identification
CellPro™ Solution Bottles offer a sterile and convenient solution for various laboratory applications. Each bottle is individually wrapped to maintain sterility and ensure reliable performance. Designed for versatility, these solution bottles are suitable for storing and dispensing various solutions used in laboratory experiments, research, and clinical settings. With their sterile packaging and robust construction, CellPro™ Solution Bottles provide peace of mind and reliability for any laboratory's needs.
Product Family Options
Product Information
- CapacitySterilityPackagingAvailabilityPrice
- 104028-186CellPro™ Reservoir bottle, roundSupplier Number: VR150Specifications:MarketsourceMarketsource
- 104028-974CellPro™ Reservoir bottle, roundSupplier Number: VR250Specifications:MarketsourceMarketsource
- 104028-944CellPro™ Reservoir bottle, roundSupplier Number: VR500Specifications:MarketsourceMarketsource
- 104028-308CellPro™ Reservoir bottle, roundSupplier Number: VR1000Specifications:MarketsourceMarketsource
- 104029-090CellPro™ Reservoir bottle, roundSupplier Number: VR2000Specifications:MarketsourceMarketsource