ContactUsLinkComponent
Upright Stainless Steel Freezer Rack for 2" Boxes, 5×5 Configuration
Upright Stainless Steel Freezer Rack for 2" Boxes, 5×5 Configuration
Catalog # 104026-834
Supplier:  Alkali Scientific
undefined
Upright Stainless Steel Freezer Rack for 2" Boxes, 5×5 Configuration
Catalog # 104026-834
Supplier:  Alkali Scientific
Supplier Number:  UFR-552
Specifications
  • Capacity:
    5×5, total 25 boxes
  • Description:
    Upright stainless steel freezer rack for 2" boxes
  • Cat. No.:
    104026-834
Specifications
About this item
Efficiently store your samples with upright stainless steel freezer rack for 2" boxes, featuring a 5×5 configuration and accommodating up to 25 boxes.
  • Constructed of corrosion resistant stainless steel
  • Racks fit standard 2" cardboard, plastic or metal boxes
  • Security lock devices available for each rack
Elevate your laboratory's sample storage capabilities with this upright stainless-steel freezer rack, designed for 2" boxes in a space-efficient 5×5 configuration. Offering a capacity of 25 boxes, this rack provides efficient organization and easy access to samples, optimizing workflow efficiency and sample management. Constructed from premium stainless steel, it ensures durability and resistance to corrosion.