ContactUsLinkComponent
Stainless Steel Freezer Racks with Drawers for 2" Boxes, 5×4 Configuration
Stainless Steel Freezer Racks with Drawers for 2" Boxes, 5×4 Configuration
Catalog # 104027-358
Supplier:  Alkali Scientific
undefined
Stainless Steel Freezer Racks with Drawers for 2" Boxes, 5×4 Configuration
Catalog # 104027-358
Supplier:  Alkali Scientific
Supplier Number:  UFDR-542
Specifications
  • Capacity:
    4×5, total 20 boxes
  • Description:
    Stainless steel freezer racks with drawers for 2" boxes
  • Cat. No.:
    104027-358
Specifications
About this item
Organize your freezer inventory with stainless steel freezer racks with drawers for 2" boxes, featuring a 5×4 configuration and holding up to 20 boxes.
  • Constructed of corrosion resistant stainless steel
  • Racks fit standard 2" cardboard, plastic or metal boxes
  • Security lock devices available for each rack
Enhance your laboratory's freezer storage with these stainless steel freezer racks featuring drawers, designed for 2" boxes in a space-saving 5×4 configuration. Offering a capacity of 20 boxes, these racks provide efficient organization and easy access to samples, streamlining sample management processes and enhancing workflow efficiency. Crafted from high-quality stainless steel, they deliver exceptional durability and resistance to corrosion, ensuring reliable performance in freezer storage environments. The inclusion of drawers adds an extra layer of protection and accessibility, making these racks indispensable for laboratories and research facilities.