ContactUsLinkComponent
Stainless Steel Freezer Racks with Drawers for 2" Boxes, 4×5 Configuration
  104028-378
undefined
Stainless Steel Freezer Racks with Drawers for 2" Boxes, 4×5 Configuration
  104028-378
 :  UFDR-452
  • Capacity:
    4×5, total 20 boxes
  • Description:
    Stainless steel freezer racks with drawers for 2" boxes
  • Cat. No.:
    104028-378
Store your samples securely with stainless steel freezer racks with drawers for 2" boxes, featuring a 4×5 configuration and accommodating up to 20 boxes.
  • Constructed of corrosion resistant stainless steel
  • Racks fit standard 2" cardboard, plastic or metal boxes
  • Security lock devices available for each rack
Optimize your freezer storage with these stainless steel freezer racks featuring drawers, designed for 2" boxes in a space-efficient 4×5 configuration. Offering a generous capacity of 20 boxes, these racks provide efficient organization and easy access to samples, enhancing workflow efficiency and sample management. Crafted from durable stainless steel, they ensure long-lasting performance and resistance to corrosion, making them suitable for use in freezer storage environments. The inclusion of drawers adds an extra layer of protection and accessibility, making these racks indispensable for laboratories and research facilities.