ContactUsLinkComponent
Stainless Steel Freezer Racks with Drawers for 2" Boxes, 4×4 Configuration
Stainless Steel Freezer Racks with Drawers for 2" Boxes, 4×4 Configuration
Catalog # 104028-540
Supplier:  Alkali Scientific
undefined
Stainless Steel Freezer Racks with Drawers for 2" Boxes, 4×4 Configuration
Catalog # 104028-540
Supplier:  Alkali Scientific
Supplier Number:  UFDR-442
Specifications
  • Capacity:
    4×4, total 16 boxes
  • Description:
    Stainless steel freezer racks with drawers for 2" boxes
  • Cat. No.:
    104028-540
Specifications
About this item
Maximize storage capacity with stainless steel freezer racks with drawers for 2" boxes, available in a 4×4 configuration and holding up to 16 boxes.
  • Constructed of corrosion resistant stainless steel
  • Racks fit standard 2" cardboard, plastic or metal boxes
  • Security lock devices available for each rack
Elevate your freezer storage capabilities with these stainless steel freezer racks featuring drawers, tailored for 2" boxes in a 4×4 configuration. Offering a generous capacity of 16 boxes, these racks provide efficient organization and easy access to samples, enhancing workflow efficiency and sample management. Constructed from durable stainless steel, they ensure long-lasting performance and resistance to corrosion, making them suitable for use in freezer storage environments. The inclusion of drawers adds an extra layer of protection and accessibility, making these racks essential for laboratory and research applications.