CellPro™ Cell Culture Flasks, TC Treated, with Plugged Caps
Supplier: Alkali Scientific
Certificates
About this item
Achieve optimal cell growth with the CellPro™ cell culture flasks, featuring TC treatment and a plugged cap for secure sealing.
- Angled neck for easy access
- Packaged in sterile, zip-sealable bags
- High clarity, 100% virgin polystyrene
- Frosted writing and clear graduations
- Notched bottom for slip free stacking
- Sterilized by E Beam
- Vented caps (with breathable membrane)
- Clear lot number for batch traceability
- Non-pyrogenic, DNase/RNase free
Elevate your cell culture experiments with CellPro™ cell culture flasks, designed to provide optimal conditions for cell growth and maintenance. These flasks are treated with tissue culture (TC) surfaces, promoting cell adhesion and proliferation, ensuring reliable and consistent results. The plugged caps ensure a secure seal, minimizing the risk of contamination and maintaining the sterility of cultures. Whether you're cultivating adherent cell lines or primary cells, these flasks offer the ideal environment for your research needs. Trust CellPro™ for premium quality and performance in cell culture applications.
Frequently Bought Together