ContactUsLinkComponent
Horizontal Head, for 4×15 ml for BenchMixer
Horizontal Head, for 4×15 ml for BenchMixer
Catalog # 104028-504
Supplier:  Alkali Scientific
undefined
Horizontal Head, for 4×15 ml for BenchMixer
Catalog # 104028-504
Supplier:  Alkali Scientific
Supplier Number:  RS7001-H150
Specifications
  • Description:
    Optional horizontal head, for 4×15 ml
  • Cat. No.:
    104028-504
Specifications
About this item
Enhance your BenchMixer with the Horizontal head for 4×15 ml tubes, providing efficient mixing for your laboratory applications.
  • Optional horizontal head for the Benchmark Mortexer Vortex Mixer RS7001
  • Hold 4×15 ml tubes
Elevate your BenchMixer with this horizontal head designed to accommodate four 15 ml tubes, optimizing mixing efficiency for a range of sample volumes. With its robust construction and precise engineering, this accessory ensures thorough and consistent mixing, ideal for various laboratory applications. Enhance your workflow and achieve reliable results with the versatility and performance of this horizontal head.