CellPro™ ASPIR-8™ Reagent Solution Reservoirs, 5ml
Supplier: Alkali Scientific
Certificates
About this item
Utilize ASPIR-8™ reagent solution reservoirs featuring a capacity of 5ml for precise dispensing of solutions.
- Specially designed for 8-Channel pipettes
- 75 mm inner chamber
- Conserve valuable reagents
ASPIR-8™ is the ideal reservoir for any laboratory utilizing 8-channel pipettes. The strict inner dimensions are designed specifically for 8-channels, thus conserving space and reagent over their longer 12-channel counterparts. This thinner design minimizes reagent exposure and usage. ASPIR-8 is the only reservoir basin that is tailor-made for your 8-channel instrument.
Frequently Bought Together