- Sterility:Sterile
- Volume:2.2 ml
- Description:96-well deep well plates
- Material:PP
- Cat. No.:104028-016
- Packaging:5/pack, 50/case
- Well shape:Square
- Bottom style:U-bottom
Maintain sterile conditions in your experiments with 2.2 ml 96-Well Deep Well Plates, featuring a U-Bottom and square well design, available in packs of 5 and cases of 50.
- Virgin, heat-resistant, medical-grade polypropylene plates
- RNase, DNase and pyrogen free
- Minimal binding and liquid retention
- Black printed alpha-numeric reference grids
- Autoclavable; sterile plates are sterilized by gamma irradiation in accordance with ISO 13485
- Temperature range: –86 to +82.2 °C
The 2.2 ml 96-Well Deep Well Plate offers researchers a sterile and reliable solution for high-throughput sample processing and storage in research laboratories. With its U-bottom and square well design, this deep well plate ensures optimal sample recovery and mixing efficiency, making it ideal for a wide range of applications, including cell culture, compound screening, and sample storage. Each pack contains 5 sterile plates, and each case contains 50 packs, providing researchers with ample supply for multiple experiments. Manufactured from premium-grade polypropylene material, these plates provide excellent chemical resistance and compatibility with various laboratory reagents and solvents. The alphanumeric labeling and raised rims facilitate easy sample identification and handling, ensuring efficient and reproducible experimental results.