Specifications
- Sterility:Non sterile
- Volume:1.0 ml
- Description:96-well deep well plates
- Material:PP
- Cat. No.:104027-170
- Packaging:5/pack, 50/case
- Well shape:Round
- Bottom style:U-bottom
Specifications
About this item
Facilitate efficient sample processing with 1.0 ml 96-Well Deep Well Plates, featuring a U-Bottom and round well design, available in packs of 5 and cases of 50.
- Virgin, heat-resistant, medical-grade polypropylene plates
- RNase, DNase and pyrogen free
- Minimal binding and liquid retention
- Black printed alpha-numeric reference grids
- Autoclavable
- Temperature range: –86 to +82.2 °C
The 1.0 ml 96-Well Deep Well Plate is designed to provide researchers with a versatile solution for high-throughput sample processing and storage in research laboratories. With its U-bottom and round well design, this deep well plate ensures optimal sample recovery and mixing efficiency, making it suitable for a wide range of applications, including cell culture, compound screening, and sample storage. Each pack contains 5 plates, and each case contains 50 packs, providing researchers with ample supply for multiple experiments. Manufactured from premium-grade polypropylene material, these plates provide excellent chemical resistance and compatibility with various laboratory reagents and solvents. The alphanumeric labeling and raised rims facilitate easy sample identification and handling, ensuring efficient and reproducible experimental results.