Specifications
- Sterility:Sterile
- Volume:1.0 ml
- Description:96-well deep well plates
- Material:PP
- Cat. No.:104028-750
- Packaging:5/pack, 50/case
- Well shape:Square
- Bottom style:U-bottom
Specifications
About this item
Maintain sterile conditions in your experiments with 1.0 ml 96-Well Deep Well Plates, featuring a U-Bottom and square well design, provided in a sterile case of 50.
- Virgin, heat-resistant, medical-grade polypropylene plates
- RNase, DNase and pyrogen free
- Minimal binding and liquid retention
- Black printed alpha-numeric reference grids
- Autoclavable; sterile plates are sterilized by gamma irradiation in accordance with ISO 13485
- Temperature range: –86 to +82.2 °C
The 1.0 ml 96-Well Deep Well Plate offers researchers a sterile and convenient solution for high-throughput sample processing and storage in research laboratories. With its U-bottom and square well design, this deep well plate ensures optimal sample recovery and mixing efficiency, making it ideal for a wide range of applications, including cell culture, compound screening, and sample storage. Manufactured from premium-grade polypropylene material, it provides excellent chemical resistance and compatibility with various laboratory reagents and solvents. Each plate features alphanumeric labeling and raised rims for easy sample identification and handling. The pack contains 50 sterile plates, ensuring the integrity of samples and reliable experimental results.