Specifications
- Sterility:Non sterile
- Volume:1.0 ml
- Description:96-well deep well plates
- Material:PP
- Cat. No.:104028-316
- Packaging:5/pack, 50/case
- Well shape:Square
- Bottom style:U-bottom
Specifications
About this item
Streamline laboratory workflows with 1.0 ml 96-Well Deep Well Plates, featuring a U-Bottom and square well design, offered in a case of 50.
- Virgin, heat-resistant, medical-grade polypropylene plates
- RNase, DNase and pyrogen free
- Minimal binding and liquid retention
- Black printed alpha-numeric reference grids
- Autoclavable
- Temperature range: –86 to +82.2 °C
The 1.0 ml 96-Well Deep Well Plate is engineered to meet the demanding requirements of high-throughput sample processing and storage in research laboratories. With its U-bottom and square well design, this deep well plate offers optimal sample recovery and mixing efficiency, making it suitable for a wide range of applications, including cell culture, compound screening, and sample storage. Manufactured from premium-grade polypropylene material, it provides excellent chemical resistance and compatibility with various laboratory reagents and solvents. Each plate features alphanumeric labeling and raised rims for easy sample identification and handling. The pack contains 50 plates, providing researchers with a reliable and cost-effective solution for high-throughput sample processing.