Specifications
- Sterility:Non sterile
- Volume:0.5 ml
- Description:96-well deep well plates
- Material:PP
- Cat. No.:104027-906
- Packaging:50/case
- Well shape:Round
- Bottom style:U-bottom
Specifications
About this item
Optimize your high-throughput screening assays with 0.5 ml 96-Well Deep Well Plates, offering a U-Bottom and round well configuration, provided in a case of 50.
- Virgin, heat-resistant, medical-grade polypropylene plates
- RNase, DNase and pyrogen free
- Minimal binding and liquid retention
- Black printed alpha-numeric reference grids
- Autoclavable
- Temperature range: –86 to +82.2 °C
The 0.5 ml 96-Well Deep Well Plate offers researchers a high-quality solution for sample storage, mixing, and processing in high-throughput applications. With its U-bottom, round well design, this deep well plate ensures optimal sample recovery and mixing efficiency, making it suitable for a wide range of laboratory procedures, including cell culture, compound screening, and sample storage. Manufactured from premium-grade polypropylene material, it provides excellent chemical resistance and compatibility with various laboratory reagents and solvents. Each plate features alphanumeric labeling and raised rims for easy sample identification and handling. The 96-Well Deep Well Plate is an essential tool for molecular biology, genomics, and proteomics research.