Order Entry
United States
ContactUsLinkComponent
4-Way Test Tube Rack
4-Way Test Tube Rack
Catalog # 104026-900
Supplier:  Alkali Scientific
undefined
4-Way Test Tube Rack
Catalog # 104026-900
Supplier:  Alkali Scientific
Supplier Number:  FR5015

Specifications

  • Description:
    4-Way tube rack, rainbow pack
  • Environmentally Preferable:
  • Cat. No.:
    104026-900
  • Packaging:
    5/pack
  • For tubes:
    4×50 ml, 12×15 ml or 32×1.5 ml/0.5 ml

Specifications

About this item

Maximize lab space with 4-way test tube rack, accommodating 4×50 ml, 12×15 ml, or 32×microtubes (0.5, 1.5, or 2 ml) in a rainbow pack.

  • 100% polypropylene
  • Designed to hold 4×50 ml, 12×15 ml, or 32×microtubes (0.5, 1.5, or 2 ml)
  • Compact and space efficient
  • Autoclavable
  • Numbered tube wells
  • Racks can be snap linked together

Elevate your laboratory workflow with 4-Way test tube rack, available in a vibrant rainbow pack. This versatile rack accommodates 4×50 ml tubes, 12×15 ml tubes, or 32×microtubes (0.5, 1.5, or 2 ml), offering flexibility to meet diverse experimental needs. Crafted from durable materials, this rack provides stability and durability for long-term use. The assorted colors not only add visual appeal but also facilitate easy sample differentiation and organization.