Orders LinkContactUsLinkComponent
CellPro™ ACK Lysing Buffer, Sterile
CellPro™ ACK Lysing Buffer, Sterile
Marketsource
Supplier:   Alkali Scientific
CellPro™ ACK Lysing Buffer, Sterile
Supplier:  Alkali Scientific
Marketsource
Certificates
CellPro™ ACK Lysing Buffer, Sterile
CellPro™ ACK Lysing Buffer, Sterile
2 Options Available Below
Product Details & Documents
Efficiently lyse red blood cells with CellPro™ ACK Lysing Buffer, ensuring reliability and sterility in your experiments.
  • For the lysis of red blood cells in samples containing white blood cells
  • Sterile
  • Filtered
CellPro™ ACK Lysing Buffer is a sterile solution designed for efficient red blood cell lysis in biological samples. This specialized buffer ensures the gentle and thorough removal of erythrocytes, facilitating downstream analysis in flow cytometry, cell sorting, and other hematological applications.
With its sterile formulation, CellPro™ ACK Lysing Buffer maintains sample integrity and minimizes the risk of contamination, ensuring reliable and reproducible results. Streamline your cell analysis workflows with the high-quality performance and convenience of CellPro™ ACK Lysing Buffer.
Product Family Options
Product Information
  • Size
    Availability
    Price
  • 104027-836
    CellPro™ ACK lysing buffer
    CellPro™ ACK lysing buffer
    100 ml
    Log in for availability
    Log in for availability
    Each
    Each - $41.48
    $41.48
    Each
       $41.48
    Minimum order quantity is 1
    Supplier Number: DB0248
    Specifications:
    Marketsource
    Marketsource
  • 104026-770
    CellPro™ ACK lysing buffer
    CellPro™ ACK lysing buffer
    1 L
    Log in for availability
    Log in for availability
    Each
    Each - $78.86
    $78.86
    Each
       $78.86
    Minimum order quantity is 1
    Supplier Number: DB0309
    Specifications:
    Marketsource
    Marketsource