1.5 ml Microcentrifuge Tubes, Clear, Conical, Lock Cap
Secure your samples with confidence using 1.5 ml clear conical microcentrifuge tubes, featuring a lock cap and free from DNase, RNase and pyrogens.
- Centrifugal speed: 29000 ×g
- Withstand boiling up to 121 °C and freezing down to –80 °C
- Snaps into place for a secure seal
- 100% polypropylene
- RNase/DNase free
- Clear, non-binding surface, graduated
- Certified RNAse, DNAse and pyrogen free
- Sterile options (SAL 10⁻⁶) and assorted colors available
The 1.5 ml microcentrifuge tubes offer clarity and convenience in laboratory settings. With a conical design and a secure lock cap, these tubes ensure reliable sample containment. They are free from DNase, RNase, and pyrogens, guaranteeing the integrity of your samples. Whether used for sample storage, centrifugation, or other applications, these tubes provide a versatile solution for your laboratory needs.
Frequently Bought Together