Specifications
- Description:3.5 ml 48-Well deep well plate
- Cat. No.:104028-282
- Packaging:5 per pack, 50 per case
- Well shape:Round
- Bottom style:U-bottom
Specifications
About this item
Enhance sample storage and processing with 3.5 ml 48-well deep well plates, featuring a U-bottom and round well design, available in a case of 50.
- Virgin, heat-resistant, medical-grade polypropylene plates
- RNase, DNase and pyrogen free
- Minimal binding and liquid retention
- Black printed alpha-numeric reference grids
- Autoclavable
- Temperature range: −86 to +82.2 °C
The 3.5 ml 48-well deep well plate is designed to meet the demanding requirements of high-throughput sample processing and storage in research laboratories. With its U-bottom, round well design, this deep well plate ensures optimal sample recovery and mixing, making it ideal for a wide range of applications, including cell culture, sample storage, and compound screening. Manufactured from high-quality polypropylene material, it offers excellent chemical resistance and compatibility with a variety of laboratory reagents and solvents. Each plate features alphanumeric labeling and raised rims for easy sample identification and handling. Sterile and RNase/DNase-free, the Alkali Scientific™ 48-Well Deep Well Plate provides researchers with a reliable platform for efficient and reproducible sample processing in molecular biology and life science applications.