ContactUsLinkComponent
Blocking Agent, Instant Nonfat Immunoblotting Grade Dry Milk
Blocking Agent, Instant Nonfat Immunoblotting Grade Dry Milk
Catalog # 104028-076
Supplier:  Alkali Scientific
CAS Number:  
undefined
Blocking Agent, Instant Nonfat Immunoblotting Grade Dry Milk
Catalog # 104028-076
Supplier:  Alkali Scientific
Supplier Number:  MB9696
CAS Number:  
Specifications
  • Description:
    Blocking agent, instant nonfat immunoblotting grade dry milk, 32 oz. jar
  • Environmentally Preferable:
  • Cat. No.:
    104028-076
Specifications
About this item
Achieve optimal blocking in immunoblotting with blocking agent, instant nonfat immunoblotting grade dry milk, providing rapid and efficient blocking.
  • Sterile
  • Filtered
The blocking agent is a premium-grade, instant nonfat dry milk powder specifically formulated for use in immunoblotting applications. Sourced from high-quality ingredients and manufactured to stringent standards, this blocking agent provides superior blocking efficiency, minimizing non-specific binding and background noise in immunoblotting experiments. With its quick-dissolving formula, it offers convenience and ease of use, allowing for rapid preparation of blocking solutions without the need for lengthy incubation times. Ideal for use in western blotting, ELISA, and other protein detection assays, the blocking agent ensures reliable and reproducible results, making it an indispensable tool for protein research laboratories.