ContactUsLinkComponent
CellPro™ Denhardt's Solution, 50X, (Frozen), Sterile Filtered
CellPro™ Denhardt's Solution, 50X, (Frozen), Sterile Filtered
  104028-982
 :  
undefined
CellPro™ Denhardt's Solution, 50X, (Frozen), Sterile Filtered
  104028-982
 :  DB0247
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    CellPro™ Denhardt's solution, 50X, (Frozen)
  • Size:
    50 ml
  • Cat. No.:
    104028-982
Ensure precise hybridization with CellPro™ Denhardt's solution, provided in 50X concentration, frozen and sterile filtered.
  • For membrane-based hybridization protocols
  • Sterile
  • Filtered
CellPro™ Denhardt's solution, a sterile and frozen 50X concentrate solution meticulously filtered for purity. With a volume of 50 ml, this solution offers convenience and efficiency for molecular biology applications. Denhardt's solution, at a concentration of 50X, provides a highly concentrated mixture of blocking agents, including Ficoll, polyvinylpyrrolidone (PVP), and bovine serum albumin (BSA), to effectively minimize non-specific binding in nucleic acid hybridization and protein blotting assays. Its sterile filtration ensures the elimination of contaminants, maintaining the integrity of your experimental results. Ideal for various molecular biology techniques, including Southern and Northern blotting, in situ hybridization, and antibody-based detection methods.