- Description:CellPro™ RIPA Buffer + Triton + Glycerol, 2X
- Size:250 ml
- Cat. No.:104027-610
Enhance protein extraction with CellPro™ RIPA Buffer + Triton + Glycerol, available in 2X concentration, sterile and filtered.
- Used to lyse cultured mammalian cells from both plated cells and cells pelleted from suspension cultures
CellPro™ RIPA Buffer + Triton + Glycerol is offered in a convenient 2X concentration, ensuring optimal performance for your protein extraction and solubilization needs. With a sterile and filtered formulation, this buffer guarantees the purity and integrity required for sensitive biochemical assays and downstream applications. RIPA buffer, combined with Triton X-100 and glycerol, facilitates efficient disruption of cells and solubilization of proteins, making it ideal for a wide range of laboratory protocols, including western blotting, immunoprecipitation and protein-protein interaction studies. The precisely calibrated composition of CellPro™ RIPA buffer ensures consistent and reliable results, empowering researchers to achieve accurate and reproducible data in their experiments.