Orders LinkContactUsLinkComponent
CellPro™ Borate Buffer, Sterile, Filtered
CellPro™ Borate Buffer, Sterile, Filtered
Marketsource
Supplier:   Alkali Scientific
CellPro™ Borate Buffer, Sterile, Filtered
Supplier:  Alkali Scientific
Marketsource
Certificates
CellPro™ Borate Buffer, Sterile, Filtered
CellPro™ Borate Buffer, Sterile, Filtered
3 Options Available Below
Product Details & Documents
Optimize your experiments with CellPro™ borate buffer, offering sterile and filtered solutions for consistent performance.
  • Colorless
  • Sterile
  • Research Grade
CellPro™ Borate buffer delivers a sterile and filtered solution essential for a wide range of laboratory procedures. Crafted with precision, this buffer solution ensures purity and reliability in scientific investigations, meeting the exacting standards of research and experimentation. The sterile nature of the buffer minimizes the risk of contamination, making it suitable for various biochemical assays, enzyme studies, and molecular biology applications. With its filtered formulation, CellPro™ borate buffer guarantees clarity and consistency in experimental outcomes, providing researchers with confidence in their results. Whether employed for protein analysis, nucleic acid electrophoresis, or other laboratory protocols, this buffer solution offers a dependable and standardized option for diverse scientific needs.
Product Family Options
Product Information
  • Size
    pH
    Availability
    Price
  • 104026-700
    CellPro™ Borate buffer, 0.5 M
    CellPro™ Borate buffer, 0.5 M
    500 ml
    8.5
    Log in for availability
    Log in for availability
    Each
    Each - $103.78
    $103.78
    Each
       $103.78
    Minimum order quantity is 1
    Supplier Number: DB0217
    Specifications:
    Marketsource
    Marketsource
  • 104028-640
    CellPro™ Borate buffered saline
    CellPro™ Borate buffered saline
    500 ml
    Log in for availability
    Log in for availability
    Each
    Each - $41.48
    $41.48
    Each
       $41.48
    Minimum order quantity is 1
    Supplier Number: DB0269
    Specifications:
    Marketsource
    Marketsource
  • 104028-024
    CellPro™ Borate buffer 0.5 M
    CellPro™ Borate buffer 0.5 M
    1 L
    9.0
    Log in for availability
    Log in for availability
    Each
    Each - $159.17
    $159.17
    Each
       $159.17
    Minimum order quantity is 1
    Supplier Number: DB0218
    Specifications:
    Marketsource
    Marketsource