Orders LinkContactUsLinkComponent
CellPro™ Flow Cytometry Tubes
CellPro™ Flow Cytometry Tubes
Marketsource
Supplier:   Alkali Scientific
CellPro™ Flow Cytometry Tubes
Supplier:  Alkali Scientific
Marketsource
Certificates
CellPro™ Flow Cytometry Tubes
CellPro™ Flow Cytometry Tubes
3 Options Available Below
Product Details & Documents
Designed for flow cytometry/FACS applications the CellPro™ flow cytometry tubes are made of clear polystyrene and are designed for fluorescence activated cell Sorting/flow cytometry equipment including FACS systems from Becton Dickinson (BD Biosciences). The diameter and wall thickness of the tube's neck is held to very tight tolerances to ensure consistent compatibility with automated instruments.
  • Compatible with FACS (BD) instruments
  • Available with 35 µm strainer cap, standard cap, or without cap
  • Clear polystyrene
  • 5 ml capacity
Product Family Options
Product Information
  • Packaging
    Sterile
    Availability
    Price
  • 104027-370
    CellPro™ Flow cytometry caps with 35 µm mesh, dual position, for 12×75 mm tubes
    CellPro™ Flow cytometry caps with 35 µm mesh, dual position, for 12×75 mm tubes
    20 Bags of 25, 500 per case
    -
    Discontinued
    Supplier Number: CT6409
    Specifications:
    Marketsource
    Marketsource
  • 104027-038
    CellPro™ Flow cytometry (FACS) tubes with standard caps, Sterile, 12×75 mm
    CellPro™ Flow cytometry (FACS) tubes with standard caps, Sterile, 12×75 mm
    25 per bag, 500 per case
    +
    Discontinued
    Supplier Number: CT6410
    Specifications:
    Marketsource
    Marketsource
  • 104028-568
    CellPro™ Flow cytometry tubes with 35 µm strainer caps, sterile, individually wrapped
    CellPro™ Flow cytometry tubes with 35 µm strainer caps, sterile, individually wrapped
    100 per case
    +
    Log in for availability
    Log in for availability
    Each
    Each - $539.94
    $539.94
    Each
       $539.94
    Minimum order quantity is 1
    Supplier Number: CT6405
    Specifications:
    Marketsource
    Marketsource
  • 104026-602
    CellPro™ Flow cytometry tubes without caps, Sterile, 12×75 mm
    CellPro™ Flow cytometry tubes without caps, Sterile, 12×75 mm
    250 per tray box, 1000 per case
    +
    Discontinued
    Supplier Number: CT6414
    Specifications:
    Marketsource
    Marketsource
  • 104027-106
    CellPro™ Standard caps for flow cytometry tubes
    CellPro™ Standard caps for flow cytometry tubes
    8 Bags of 125 caps, 1000 per case
    -
    Log in for availability
    Log in for availability
    Each
    Each - $103.78
    $103.78
    Each
       $103.78
    Minimum order quantity is 1
    Supplier Number: CT6419
    Specifications:
    Marketsource
    Marketsource
  • 104028-366
    CellPro™ Flow cytometry tubes without caps, bulk econo-pack
    CellPro™ Flow cytometry tubes without caps, bulk econo-pack
    4000 per case
    -
    Log in for availability
    Log in for availability
    Each
    Each - $346.09
    $346.09
    Each
       $346.09
    Minimum order quantity is 1
    Supplier Number: CT6420
    Specifications:
    Marketsource
    Marketsource