SureSeal™ Sterile Transparent Mailing Tubes
Supplier: Alkali Scientific
Certificates
About this item
Safely transport and store samples with SureSeal™ sterile transparent mailing tubes, ensuring integrity throughout transit.
- Leak-proof, 1.25 turn screw-cap
- Meet 95 kPa transportation guidelines
- Vivid, printed graduations
- Screw-caps attached
- Sterile, RNase, DNase, pyrogen free
Transport tubes, sometimes called mailing tubes, are used in multiple sectors of the laboratory market, including medical, hospital, environmental, independent testing, food testing and biological research laboratories. Used to store, transport or freeze samples, these tubes come in both 5 ml (16×60 mm) and 10 ml (16×80 mm) and are fitted with leak-proof screw-caps. They are made of high-grade polypropylene, and include white printed graduations and marking area. Tubes are self-standing.
Packaging: Packaged in 10 bags 100 tubes.
Frequently Bought Together