2-Tiered Rack For 5 ml Macrotubes , 16 Place, 2 Per Pack
Catalog # 104029-018
Supplier: Alkali Scientific
Specifications
- Description:Two-tiered tube rack for 16×5 ml MacroTube®
- Cat. No.:104029-018
Specifications
About this item
Maximize bench space efficiency with 2-tiered rack for 5 ml macrotubes, accommodating 16 tubes in a pack of 2.
- Designed specifically for 5 ml macrotubes and Eppendorf tubes
- Accepts 5 ml snap cap and 5 ml screw cap tubes
A great workstation tube rack for 16×5 ml MacroTube, package of 2 screw cap tubes, designed specifically for 5 ml MacroTube ® (also accepts Eppendorf). Tubes are held securely in custom shaped wells that accepts 5 ml snap cap and 5 ml screw cap tubes. Workstation is also perfect for 15 ml conical bottom tubes.
Two tiers make it easier to pipettetete into the second row and easier to see which tubes are filled or empty.