Order Entry
United States
ContactUsLinkComponent
Cryogenic Boxes with Hinged Lid
Cryogenic Boxes with Hinged Lid
Catalog # 104027-052
Supplier:  Alkali Scientific
undefined
Cryogenic Boxes with Hinged Lid
Catalog # 104027-052
Supplier:  Alkali Scientific
Supplier Number:  C2581

Specifications

  • Description:
    Cryostorage box, 25 place (5×5), Polycarbonate, with pull-off lid, 5 per pack
  • Dimensions:
    5.25×5.25×2.75"
  • Cat. No.:
    104027-052

Specifications

About this item

Organize and protect your cryogenic samples with cryogenic boxes featuring a hinged lid.

  • Polycarbonate, withstand −196 °C
  • Convenient hinged lid
  • Includes drainage slots for LN2

Ultra-low temperature storage boxes are designed to hold 1.2 to 2.0 ml cryogenic storage vials.

An excellent and long-lasting replacement for cardboard boxes, they are fully autoclavable and withstand temperatures ranging from −196 to +121 °C. they are compatible with all standard 133 mm (5.25") freezer racks.

Clear lids are hinged and have printed indexing for sample identification purposes.