ContactUsLinkComponent
Upright Stainless Steel Freezer Rack for Standard 2" Boxes, 3×5" Configuration
Upright Stainless Steel Freezer Rack for Standard 2" Boxes, 3×5" Configuration
  104028-816
undefined
Upright Stainless Steel Freezer Rack for Standard 2" Boxes, 3×5" Configuration
  104028-816
 :  UF352
  • Capacity:
    3×5, total 15 boxes
  • Description:
    Upright stainless steel freezer rack for standard 2" boxes
  • Cat. No.:
    104028-816
Maximize freezer space with the upright stainless steel freezer rack for standard 2" boxes, offering a box capacity of 3" deep and 5" tall, accommodating up to 15 boxes.
  • Constructed of corrosion resistant stainless steel
  • Racks fit standard 2" cardboard, plastic or metal boxes
  • Security lock devices available for each rack
Elevate your freezer storage with this upright stainless steel freezer rack, meticulously engineered to accommodate standard 2" boxes in a versatile configuration. With a capacity of 15 boxes arranged in a layout of 3" deep and 5" tall, this rack offers ample storage space while optimizing accessibility to samples. Crafted from high-quality stainless steel, it ensures durability and stability, providing reliable support for long-term sample preservation. Whether used for sample organization or inventory management, this freezer rack is an indispensable asset in any laboratory setting.