- Cap type:Vented cap
- Sterility:Sterile
- Description:5 Layer, cell culture multilayer flask, TC treated
- Packed:1/pack, 8/case
- Cat. No.:104027-288
- Culture area:870 cm²
Optimize cell culture conditions with the 5-layer cell culture flask, providing a multi-layered design for increased culture capacity and efficiency.
- Vacuum plasma TC treatment for the best cell attachment
- Sterilized by E-Beam
- Non-pyrogenic, DNase/RNase free
- Vented cap
The 5-layer cell culture flask represents a pinnacle of innovation in cell culture technology, meticulously designed to cater to the diverse needs of modern laboratory settings. Its sophisticated multi-layered construction not only facilitates superior gas exchange, ensuring an optimal growth environment for cells, but also serves as a robust barrier against contaminants, safeguarding the integrity of your cultures.
Crafted from premium materials, this flask boasts exceptional durability and reliability, instilling confidence in researchers as they embark on their scientific endeavors. Its ergonomic form factor and seamlessly fitting lid enhance user experience, streamlining workflows and promoting efficiency in the laboratory. Whether you're conducting routine cell culture experiments or pursuing groundbreaking research, the 5-Layer cell culture fask stands as a testament to precision engineering and unparalleled performance, empowering scientists to push the boundaries of discovery with confidence and ease.