Order Entry
United States
ContactUsLinkComponent
TRAzol® RNA Purification Reagent Kit
TRAzol® RNA Purification Reagent Kit
Catalog # 104028-922
Supplier:  Alkali Scientific
CAS Number:  
undefined
TRAzol® RNA Purification Reagent Kit
Catalog # 104028-922
Supplier:  Alkali Scientific
Supplier Number:  TRZ-100
CAS Number:  

Specifications

  • Description:
    TRAzol® RNA purification reagent kit, 100 ml, store at 2 - 8 °C, protect from Light
  • Cat. No.:
    104028-922

Specifications

About this item

Isolate high-quality RNA efficiently with the TRAzol® RNA purification reagent kit, available in a 100 ml size for versatile research applications.

  • Permits the isolation of RNA, DNA, and protein from the same sample
  • Offers superior lysis capability, even with difficult sample types
  • Optimized formulations and protocols for tissues, cells, serum, virus and bacteria
  • Reliably purify RNA from multiple sample volumes and sources
  • Performs well with small quantities of tissue (50 to 100 mg) and cells (5×106), as well as with large quantities of tissue (≥1 g) and cells (>107)
  • Comes with protocols for purification from samples of human, animal, plant or bacterial origin
  • Store at 4 °C, protect from light
  • Stable for up to 12 months when properly stored
  • RNA extraction method and characteristics identical to TRAzol®

TRAzol® RNA purification reagent kit, 100 ml TRAzol® Reagent is a complete, ready-to-use reagent for the isolation of high-quality total RNA or the simultaneous isolation of RNA, DNA, and protein from a variety of biological samples. This monophasic solution of phenol and guanidine isothiocyanate is designed to isolate separate fractions of RNA, DNA, and proteins from cell and tissue samples of human, animal, plant, yeast, or bacterial origin, within one hour.

Key features of TRAzol® reagent include:
• Permits the isolation of RNA, DNA, and protein from the same sample
• Offers superior lysis capability, even with difficult sample types
• Optimized formulations and protocols for tissues, cells, serum, virus and bacteria

Reliably purify RNA from multiple sample volumes and sources TRAzol® reagent performs well with small quantities of tissue (50 to 100 mg) and cells (5×106), as well as with large quantities of tissue (≥1 g) and cells (>107), and comes with protocols for purification from samples of human, animal, plant, or bacterial origin. TRAzol® Reagent maintains the integrity of the RNA due to highly effective inhibition of RNase activity while disrupting cells and dissolving cell components during sample homogenization. The simplicity of the TRAzol® Reagent method allows simultaneous processing of a large number of samples. The entire procedure can be completed in 1 hour.

Total RNA isolated by TRAzol® Reagent is free of protein and DNA contamination. Formulated for isolation of multiple molecular targets TRAzol® Reagent allows you to perform sequential precipitation of RNA, DNA, and proteins from a single sample. After homogenizing the sample with TRAzol® Reagent, chloroform is added, and the homogenate is allowed to separate into a clear upper aqueous layer (containing RNA), and interphase and red lower organic layers (containing the DNA and proteins). RNA is precipitated from the aqueous layer with isopropanol. DNA is precipitated from the interphase/organic layer with ethanol. Protein is precipitated from the phenol-ethanol supernatant by isopropanol precipitation. The precipitated RNA, DNA, or protein is washed to remove impurities, and then resuspended for use in downstream applications.

Sample Type (General): Bacteria, yeast, blood, tissue, viral samples, plant samples, cells high throughput compatibility: Not high throughput-compatible (manual) downstream.
Application: Reverse Transcriptase PCR (RT-PCR), real-time Quantitative PCR (qPCR), nuclease protection assays, cDNA Library construction, northern blotting.
Cloning scale: Scalable system: TRAzol®.
Quantity: 1 Product size: 100 ml.
Final Product: Total RNA, micro RNA, Transcriptome RNA.
Purification time: 1 hr.
Number of reactions: 100 Preps.
Isolation Technology: Organic extraction.
Starting material (Amount): Up to 1×10⁻⁷ cells, Up to 1 g tissue.
Shipping condition: Room temperature contents and storage One bottle. Store at room temperature.