Tip Combs, 96, V-Bottom, PP, for KingFisher™ and IsoPURE™
Catalog # 104027-266
Supplier: Alkali Scientific
Specifications
- Description:Tip comb, 96, PP, for KingFisher™ and IsoPURE™
- Environmentally Preferable:
- Cat. No.:104027-266
- Packaging:50 bags of 2 pieces, 100/case
- Bottom style:V-bottom
Specifications
About this item
Facilitate automated liquid handling with the tip comb, designed for 96 V-bottom wells, made of polypropylene, compatible with KingFisher™ and IsoPURE™ systems, supplied in 50 bags of 2 pieces, and packaged in a case of 100.
- Compatible with KingFisher™ Flex
- Low-binding, medical-grade polypropylene
- RNase, DNase free
The tip comb designed for use with KingFisher™ and IsoPURE™ systems. Each comb features 96 tips with V-bottom geometry, allowing efficient liquid handling and sample extraction. Manufactured from polypropylene (PP), these tip combs ensure compatibility with a wide range of chemical reagents and solvents, making them suitable for various laboratory applications. Tip combs to enhance the efficiency and productivity of your automated liquid handling systems.
Packaging: Packaged in convenient bags containing two pieces each, with 50 bags per case, these tip combs offer flexibility and ease of use in high-throughput workflows.