Order Entry
United States
Orders LinkContactUsLinkComponent
CellPro™ DPBS Buffer, 1X

CellPro™ DPBS Buffer, 1X

CellPro™ DPBS buffer, 1X, 500 ml
Catalog # 104028-846
Marketsource
Supplier:   Alkali Scientific
Supplier #:  TBD555
CellPro™ DPBS Buffer, 1X
Catalog # 104028-846
Supplier:  Alkali Scientific
Marketsource
CAS Number:  
CellPro™ DPBS Buffer, 1X
CellPro™ DPBS Buffer, 1X

CellPro™ DPBS buffer, 1X, 500 ml

CAS Number:  
Price
$16.55
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Maintain cell viability with CellPro™ DPBS buffer, 1X, available in a 500 ml bottle for cell culture experiments.

  • For molecular biology
  • 0.2 μm filtered
  • Sterile
  • Ultra Pure

CellPro™ DPBS Buffer, offered in a 1X concentration and a convenient 500 ml volume, provides a balanced salt solution essential for numerous cell culture and molecular biology procedures. Its molecular biology grade ensures high purity and reliability in maintaining cellular health and integrity during experiments.

Specifications

Specifications

  • Description:
    CellPro™ DPBS buffer, 1X, 500 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.