ContactUsLinkComponent
In situ Adapter for Use with Combination Blocks for Thermal Cycler
In situ Adapter for Use with Combination Blocks for Thermal Cycler
Catalog # 104028-062
Supplier:  Alkali Scientific
undefined
In situ Adapter for Use with Combination Blocks for Thermal Cycler
Catalog # 104028-062
Supplier:  Alkali Scientific
Supplier Number:  T5000-A-IS
Specifications
  • Description:
    In-Situ adapter for use with combination blocks
  • Cat. No.:
    104028-062
Specifications
About this item
Enhance thermal cycling experiments with the in situ adapter, designed for use with combination blocks in your thermal cycler.
  • In situ adapter fits on the TC 9639 thermal cycler block and holds four standard slides
This specialized adapter, compatible with combination blocks for thermal cyclers, facilitates precise and efficient in situ procedures. Designed for seamless integration with your existing equipment, it ensures reliable and reproducible results in molecular biology applications.