12-Strip Tubes and 12-Strip Flat Caps Packed Together
104027-398
- Description:12-strip tubes and 12-strip flat caps packed together, 80 strips/pack, 960 tubes and caps
- Cat. No.:104027-398
Simplify laboratory workflow with the 12-strip tubes and flat caps combo pack. This convenient pack includes 80 strips, each containing 12 tubes and 12 flat caps providing you with a total of 960 tubes and caps.
- Angularly attached caps prevent cap lids and hinges from interfering with each other, allowing for easier operation in an SBS format rack
- Ultra-thin walls ensure efficient thermal transfer and maximum yield, yet are strong enough to prevent crushing
- Clear tubes provide high clarity for visualizing contents
- For use in pharmaceutical industry, food industry, and molecular biology
- Manufactured from high-quality, virgin polypropylene to ensure purity and provide excellent optical clarity
- Certified free of detectable RNase, DNase, DNA, PCR inhibitors and tested pyrogen-free
Designed for compatibility with standard PCR machines, these tubes and caps ensure secure sealing and reliable sample containment during PCR amplification. The attached flat caps offer easy handling and minimize the risk of cross-contamination between samples.