ContactUsLinkComponent
ProCycle™ PCR Tubes and Separate Caps Combo, 0.2 ml, 12-Strip
ProCycle™ PCR Tubes and Separate Caps Combo, 0.2 ml, 12-Strip
Catalog # 104027-570
Supplier:  Alkali Scientific
undefined
ProCycle™ PCR Tubes and Separate Caps Combo, 0.2 ml, 12-Strip
Catalog # 104027-570
Supplier:  Alkali Scientific
Supplier Number:  PC7012
Specifications
  • Description:
    12 strip PCR tubes and 12 strip dome caps combo, 0.2 ml, 80 strips of 12 tubes/caps, 960/pack
  • Cat. No.:
    104027-570
Specifications
About this item
Streamline PCR workflows with ProCycle™ 0.2 ml 12-srip PCR tubes and separate caps combo pack, containing 12-strip tubes and 12-strip flat caps packed together.
  • Ultra-thin walls ensure efficient thermal transfer and maximum yield, yet are strong enough to prevent crushing
  • Clear tubes provide high clarity for visualizing contents
  • For use in pharmaceutical industry, food industry and molecular biology
  • Manufactured from high-quality, virgin polypropylene to ensure purity and provide excellent optical clarity
  • Certified free of detectable RNase, DNase, DNA, PCR inhibitors and tested pyrogen-free
Streamline your PCR setup with ProCycle™ 0.2 ml 12-Strip PCR tubes and separate caps combo pack. This convenient kit includes 12-strip PCR tubes and separate flat caps, offering flexibility and ease of use in PCR applications. Each strip contains 12 individual tubes, pre-attached with flat caps to prevent sample evaporation and contamination. With a total of 80 strips per pack, this combo kit provides ample supply for multiple PCR experiments. ProCycle™ PCR tubes and caps for reliable performance and convenience in your molecular biology workflows.