ContactUsLinkComponent
Flexible Sealing Mat, Reuseable
Flexible Sealing Mat, Reuseable
Catalog # 104026-578
Supplier:  Alkali Scientific
undefined
Flexible Sealing Mat, Reuseable
Catalog # 104026-578
Supplier:  Alkali Scientific
Supplier Number:  PC50
Specifications
  • Description:
    Flexible sealing mat for any 96 well PCR plates, reuseable
  • Cat. No.:
    104026-578
  • Packaging:
    10/pack
Specifications
About this item
Ensure a tight seal for PCR plates, reusable.
  • Flexible sealing mat for any 96 well PCR plates
Ensure reliable sealing and contamination protection for 96-well PCR plates with flexible sealing mats. These reusable mats are compatible with any standard 96-well PCR plate, providing a secure seal to prevent sample evaporation and cross-contamination during PCR cycling. Crafted from high-quality materials, these mats offer excellent chemical resistance and durability, allowing for repeated use without compromising performance. Each pack contains 10 sealing mats, offering cost-effective sealing solutions for your laboratory's PCR workflows.