ContactUsLinkComponent
OmniPette™ Single Channel Pipette
OmniPette™ Single Channel Pipette
Catalog # 104026-792
Supplier:  Alkali Scientific
undefined
OmniPette™ Single Channel Pipette
Catalog # 104026-792
Supplier:  Alkali Scientific
Supplier Number:  P9280-50U
Specifications
  • Volume:
    50 µl
  • Description:
    OmniPette™ single channel pipette
  • Cat. No.:
    104026-792
Specifications
About this item
Achieve accurate dispensing with OmniPette single channel pipette, offering a volume range of 5 to 50 μl.
  • Stainless steel pistons and teflon seals
  • Superior accuracy and reproducibility
  • Adjustable volume and locking wheel
  • Full volume range (0.1 µl to 10 ml)
  • Outstanding user comfort
  • Fully autoclavable
  • 3 year warranty
Experience unparalleled precision and versatility with the OmniPette™ Single channel pipette. This pipette offers exceptional performance for a wide range of liquid handling tasks in the laboratory. Its ergonomic design ensures comfortable operation during extended use, while the adjustable volume setting allows precise dispensing of samples with ease and confidence. OmniPette™ Single channel pipette for accurate and reliable pipetting results in your research projects.