12-Channel Mechanical Pipettes
Supplier: Alkali Scientific
Certificates
About this item
Efficiently handle multiple samples with 12-channel mechanical pipettes, ideal for high-throughput applications.
- Precision designed tip cones provide leak-proof tip fit
- Continuously adjustable volume selection with thumbwheel or push-button
- Curved ejector bar reduces ejection force
- Fully autoclavable
- Large Capacity Multichannel Pipette
Experience unparalleled efficiency and precision in liquid handling tasks with 12-channel mechanical pipettors. Engineered to streamline pipetting workflows, these pipettors enable simultaneous aspiration and dispensing across multiple wells or tubes, maximizing productivity in laboratory settings. With adjustable volume settings and ergonomic design, they ensure comfortable and accurate pipetting, even during high-throughput applications.
Frequently Bought Together