CellPro™ Cryo Storage Box, 100 Place, 10×10, Blue, PC with Clear Hinged Lid, without Barcode
Catalog # 104027-162
Supplier: Alkali Scientific
Specifications
- Color:Blue
- Description:Cryostorage box, 100 place, 10×10, with clear hinged lid, without barcode
- Material:Polycarbonate (PC)
- Cat. No.:104027-162
Specifications
About this item
Store samples securely with cryo storage box, featuring 100 places, blue PC construction with a clear hinged lid.
- Polycarbonate, withstands −196 °C
- Convenient hinged lid
- Includes drainage slots for LN₂
Ensure the safe storage of cryogenic samples with the cryo storage box. Featuring 100 individual compartments arranged in a 10×10 grid, this box is constructed from durable polycarbonate in a striking blue color. The clear hinged lid provides visibility and protection for your samples, while the absence of a barcode allows flexible labeling options. Each pack contains five cryo storage boxes, offering a reliable solution for sample organization and preservation in your laboratory.