CellPro™ Cryostorage Box, 100 place (10×10), Polycarbonate, with Hinged Lid
104027-166
- Description:Cryostorage box, 100 place (10×10), with winged lid (5.25×5.25×2"), 5/pack
- Material:Polycarbonate (PC)
- Cat. No.:104027-166
Organize cryostorage with cryostorage box, featuring 100 places (10×10), polycarbonate construction, and a hinged lid.
- 10×10 Format
- 100% Polycarbonate
Organize and safeguard cryogenic samples with the cryostorage box. Designed for efficient storage of 100 vials or tubes in a 10×10 grid configuration, this box is made from durable polycarbonate for long-lasting performance. The hinged lid ensures secure closure, while the compact size (5.25×5.25×2") allows space-saving storage in freezers or liquid nitrogen tanks. Each pack contains five cryostorage boxes, providing a reliable solution for sample preservation in your laboratory.