Locking Clips, with Breakaway Lifting Tabs
Catalog # 104029-088
Supplier: Alkali Scientific
Specifications
- Description:Stop-Pop™ locking clips, with breakaway lifting tabs for 1.5 ml tubes, 100/pack
- Cat. No.:104029-088
Specifications
About this item
Secure tubes with ease using Stop-Pop™ Locking clips, designed for 1.5 to 2 ml tubes.
- Prevents 1.5 to 2 ml minicentrifuge tubes from popping open during heating/boiling
Secure sample tubes with ease using the Stop-Pop™ Locking clips. These locking clips feature breakaway lifting tabs for convenient tube retrieval. The innovative design ensures a tight seal to prevent sample leakage or contamination during storage or transport. Each pack contains 100 locking clips, providing an efficient solution for tube closure in laboratory workflows.