CellPro™ U-Bottom, TC-Treated Cell Culture Plates with Lids
104028-666
- No. of wells:96
- Description:Cell culture plates with lids, TC treated
- Material:PS
- Environmentally Preferable:
- Cat. No.:104028-666
- Packaging:100 per pack
- Bottom style:Round bottom
Conduct cell culture experiments with precision using the Alkali Scientific™ CellPro™ U-bottom cell culture plates, featuring TC treatment and supplied with lids for convenience.
- High clarity, 100% virgin polystyrene
- Vacuum gas plasma tissue culture treatment
- Clear lot number for batch tractability
- Peel-to-open, individually wrapped and sterilized by ga mma irradiation
- Markings of well coordinates available for 12, 24, 48 and 96 well plates
- Non-pyrogenic, DNase/RNase free
- Gamma sterilized
Elevate your cell culture experiments with CellPro™ U-bottom, TC-treated cell culture plates with lids. Crafted with precision and quality in mind, these plates offer a superior surface treatment that enhances cell adhesion and growth, ensuring reliable and consistent results. The U-bottom design promotes efficient sample recovery and minimizes dead volume, optimizing the performance of your assays. Each plate is equipped with a secure-fitting lid, maintaining sterility and preventing contamination throughout your experiments.