Specifications
- No. of wells:96
- Sterility:Sterile
- Description:Cell culture plates with lids, non-treated
- Material:PS
- Cat. No.:104028-956
- Packaging:100 per case
- Bottom style:U-bottom
Specifications
About this item
Grow cell cultures effectively in CellPro™ U-bottom, non-treated cell culture plates, featuring lids to maintain sterility and prevent contamination.
- High clarity, 100% virgin polystyrene
- Vacuum gas plasma tissue culture treatment
- Clear lot number for batch tractability
- Peel-to-open, individually wrapped and sterilized by ga mma irradiation
- Markings of well coordinates available for 12, 24, 48 and 96 well plates
- Non-pyrogenic, DNase/RNase free
- Gamma sterilized
Alkali Scientific™ introduces CellPro™ U-bottom cell culture plates, designed to provide optimal conditions for cell growth and experimentation. Featuring a U-bottom design, these plates promote uniform cell distribution and facilitate efficient media aspiration and sample recovery. Each plate comes with a matching lid to maintain sterility and prevent contamination during long-term cultures. Made from high-quality materials, these non-treated plates offer excellent optical clarity for microscopy and imaging applications. Whether you're conducting cell-based assays, drug screening, or tissue culture experiments, trust CellPro™ U-bottom cell culture plates for reliable performance and reproducible results.