ProCycle™ Optical qPCR Sealing Film
104028-224
- Description:Real-Time qPCR sealing film, adhesive
- Cat. No.:104028-224
- Packaging:Quantity 100
Seal qPCR plates effectively with ProCycle™ Optical qPCR sealing film, available in a pack of 100 for reliable performance.
- Polyolefin film
- Certified RNAse, DNAse and DNA-free
- Functional from –40 to +120 °C
With the highest transparency, this polyolefin film offers a special pressure sensitive DMSO-resistant silicone adhesive embedded inside the film itself. A polyester backing with end tabs assures easy positioning of the plate. This film is also perfectly suited for raised-rim plates ensuring reliable sealing around each well. Certified RNAse, DNAse and DNA-free.