ContactUsLinkComponent
Petri Dish, 150×15 mm, Sterile, Stackable
Catalog # 104026-598
Supplier:  Alkali Scientific
undefined
Petri Dish, 150×15 mm, Sterile, Stackable
Catalog # 104026-598
Supplier:  Alkali Scientific
Supplier Number:  PD3572
Specifications
  • Description:
    Petri dish, 150×15 mm, sterile, stackable, 10 per pack
  • Material:
    PS
  • Cat. No.:
    104026-598
  • Packaging:
    100 case
Specifications
About this item
Ensure reliable culture growth with Alkali Scientific™ Petri Dish, sized at 150×15 mm and provided in sterile packaging.
  • High clarity polystyrene
  • Flat, transparent surface
  • Stackable for easy storage and handling
  • Sterilized by E-Beam
  • Non-pyrogenic
Maximize efficiency in your microbiology lab with our Petri Dish, 150×15 mm. These sterile dishes are stackable, optimizing space utilization and facilitating organized culture setups. Designed for reliability and convenience, these dishes are ideal for a wide range of microbiological applications, from bacterial culture to antibiotic susceptibility testing. Trust in the quality and performance of our Petri Dishes to support your research endeavors. These non-treated polystyrene plates are transparent and DNase/RNase-Free. The bottom has a ridge allowing for easy handling and stacking. The plate has three vents for gas exchange and compartments for bacterial culture use with different media under the same conditions.
Packaging: 10 dishes per pack and 100 cases.