ContactUsLinkComponent
Petri Dish 100×15 mm, Sterile, Standard, Stackable, Semi Slippable
Petri Dish 100×15 mm, Sterile, Standard, Stackable, Semi Slippable
Catalog # 104027-058
Supplier:  Alkali Scientific
undefined
Petri Dish 100×15 mm, Sterile, Standard, Stackable, Semi Slippable
Catalog # 104027-058
Supplier:  Alkali Scientific
Supplier Number:  PD3571
Specifications
  • Description:
    Petri dish 100×15 mm, sterile, standard, stackable, semi slippable, 20 per inner bag
  • Material:
    PS
  • Cat. No.:
    104027-058
  • Packaging:
    500 case
Specifications
About this item
Maintain sterile conditions with Alkali Scientific™ Petri Dish 100×15 mm, featuring a standard design, stackable configuration, and semi slippable surface.
  • High clarity polystyrene
  • Flat, transparent surface
  • Stackable for easy storage and handling
  • Sterilized by E-Beam
  • Non-pyrogenic
Experience unparalleled convenience and reliability with our Petri Dish 100×15 mm. Sterile and stackable, these dishes offer a standardized solution for culturing microorganisms in laboratory settings. With semi-slippable surfaces, they provide ease of handling while minimizing the risk of accidental movement. These non-treated polystyrene plates are transparent and DNase/RNase-Free. The bottom has a ridge allowing for easy handling and stacking. The plate has three vents for gas exchange and compartments for bacterial culture use with different media under the same conditions.
Packaging: Packaged with 20 dishes per inner bag and 500 cases.