ProCycle™ 0.1 ml Bio-Rad Type 96 Well PCR Plates, without Skirt
Catalog # 104027-116
Supplier: Alkali Scientific
Specifications
- No. of wells:96
- Skirt:Non-Skirted
- Well volume:0.1 ml
- Description:Bio-Rad type PCR plates, white, 25 per pack, 100 per case
- Cat. No.:104027-116
Specifications
About this item
Conduct experiments with precision using Alkali Scientific™ ProCycle™ 0.1 ml Bio-Rad type 96 well PCR Plates, featuring no skirt design in white.
- Easy wellidentification: High-contrast, black alphanumeric indexing facilitates quick and accurate well identification, streamlining workflow
- Optimal heat transfer: Ultra-thin, uniform wells ensure optimal heat transfer, promoting high reaction efficiency and reproducibility
- Sample protection: Well ridges help reduce sample evaporation when sealing with film, foil or strip caps, preserving sample integrity
- High clarity, Ultra-pure material: PCR plate is manufactured from high-clarity, ultra-pure polypropylene, meeting FDA 21 CFR 177.1520 and USP Class VI standards
- Purity assurance: Plate is certified free of detectable DNase, RNase, DNA, PCR inhibitors and has been tested to be pyrogen-free
- Nearly identical to: Bio-Rad item mlL9601 and additionally Thermo Scientific item AB-0700 plate. Direct substitutes
Facilitate your PCR experiments with ProCycle™ 0.1 ml Bio-Rad type 96 well PCR Plates. Featuring a no-skirt design and white color for enhanced contrast, these plates offer optimal sample visibility and easy well identification. Compatible with a range of PCR instruments, these plates are essential tools for your molecular biology workflows.
Packaging: Each pack contains 20 plates, and each case includes 100 plates, ensuring an abundant supply for your laboratory needs.