ProCycle™ 0.1 ml Bio-Rad CFX/Thermo Type 96 Well PCR Plate, Low Profile, without Skirt
Supplier: Alkali Scientific
Certificates
About this item
Perform low-profile PCR assays with Alkali Scientific™ ProCycle™ 0.1 ml Bio-Rad CFX/Thermo type 96 well PCR Plate, featuring no skirt design and clear color.
- Easy well identification: High-contrast, black alphanumeric indexing facilitates quick and accurate well identification, streamlining workflow
- Optimal heat transfer: Ultra-thin, uniform wells ensure optimal heat transfer, promoting high reaction efficiency and reproducibility
- Sample protection: Well ridges help reduce sample evaporation when sealing with film, foil or strip caps, preserving sample integrity
- High clarity, Ultra-pure material: PCR plate is manufactured from high-clarity, ultra-pure polypropylene, meeting FDA 21 CFR 177.1520 and USP Class VI standards
- Purity assurance: Plate is certified free of detectable DNase, RNase, DNA, PCR inhibitors and has been tested to be pyrogen-free
- Nearly identical to: Bio-Rad item mlL9601 and additionally Thermo Scientific item AB-0700 plate. Direct substitutes
Strea mline your PCR workflow with ProCycle™ 0.1 ml Bio-Rad CFX/Thermo type 96 Well PCR Plates. These low-profile plates feature a no-skirt design for compatibility with various PCR instruments, including Bio-Rad CFX and Thermo models. With clear color for optimal sample visibility and easy identification, these plates are ideal for high-throughput PCR applications.
Packaging: Each pack contains 20 plates, and each case includes 100 plates, providing an ample supply for your molecular biology experiments.
Frequently Bought Together