ContactUsLinkComponent
ProCycle™ Flat 8-Strip PCR Tube Caps
ProCycle™ Flat 8-Strip PCR Tube Caps
Catalog # 104028-928
Supplier:  Alkali Scientific
undefined
ProCycle™ Flat 8-Strip PCR Tube Caps
Catalog # 104028-928
Supplier:  Alkali Scientific
Supplier Number:  PCN7056
Specifications
  • Color:
    Clear
  • Description:
    Flat caps for 8-Strip PCR tubes
  • Cat. No.:
    104028-928
  • Packaging:
    125 per pack, 1250 per case
Specifications
About this item
Seal PCR tubes securely with Alkali Scientific™ ProCycle™ Flat 8-Strip PCR tube caps, available in packs of 125 and cases of 1250.
  • Exceptional thermal conductivity for precise thermal cycling
  • Optically clear
Seal PCR reactions securely with ProCycle™ flat 8-Strip PCR tube caps. These caps provide a tight and leak-proof seal to prevent sample evaporation and contamination during PCR amplification. With a flat design for convenient storage and handling, the caps ensure reliable sample protection while maintaining ease of use.
Packaging: Each pack contains 125 caps, and each case includes 1250 caps, providing abundant supply for your laboratory needs.