ProCycle™ Dome Caps for 8-PCR Strip Tubes
104028-558
- Color:Clear
- Description:8-Strip dome caps for PCR tubes
- Cat. No.:104028-558
- Packaging:125 pack
Securely seal PCR strip tubes with Alkali Scientific™ ProCycle™ dome caps, available in packs of 125 for convenient use.
- Exceptional thermal conductivity for precise thermal cycling
- Optically clear
Ensure reliable sealing and protection of PCR strip tubes with ProCycle™ dome caps. These caps provide a secure and leak-proof seal, preventing sample evaporation and contamination during PCR amplification. Featuring a domed design that conforms to the shape of the PCR tubes, these caps ensure a tight fit and enhanced sealing performance. Designed for convenience and ease of use, the dome caps offer reliable sample protection, maintaining sample integrity throughout the PCR process.