ProCycle™ Half-Skirted PCR Plates, 96×0.2 ml Well, Black Printer Alphanumerics, A12 notch
104027-740
- No. of wells:96
- Description:Mini skirted PCR plates, half-skirted, 96×0.2 ml well, black printed alphanumerics, A12 cut corner, 10 per pack
- Cat. No.:104027-740
Streamline your PCR workflow with Alkali Scientific™ ProCycle™ half-skirted PCR plates, featuring 96×0.2 ml well, black printed Alphanumerics, and an A12 notch.
- Universal Compatibility: These PCR plates are compatible with a broad range of thermal cyclers, ensuring versatility and adaptability in your lab
- Easy well identification: High-contrast, black alphanumeric indexing facilitates quick and accurate well identification, streamlining workflow
- Optimal heat transfer: Ultra-thin, uniform wells ensure optimal heat transfer, promoting high reaction efficiency and reproducibility
- Sample protection: Well ridges help reduce sample evaporation when sealing with film, foil or strip caps, preserving sample integrity
- High clarity, ultra-pure material: PCR plate is manufactured from high-clarity, ultra-pure polypropylene, meeting FDA 21 CFR 177.1520 and USP Class VI standards
- Purity assurance: The plate is certified free of detectable DNase, RNase, DNA, PCR inhibitors and has been tested to be pyrogen-free
Compatible with the following machines:
• ABI 7000, ABI 7300, ABI 7500, ABI 7700 and ABI StepOne
• BioRad iCycler, BioRad myIQ and BioRad myIQ5
• ABI Sequencer 3100 and ABI Sequencer 3132