Order Entry
United States
ContactUsLinkComponent
ProCycle™ 0.2 ml 96 Well PCR Plates, Semi Skirt, White
ProCycle™ 0.2 ml 96 Well PCR Plates, Semi Skirt, White
Catalog # 104027-780
Supplier:  Alkali Scientific
undefined
ProCycle™ 0.2 ml 96 Well PCR Plates, Semi Skirt, White
Catalog # 104027-780
Supplier:  Alkali Scientific
Supplier Number:  PCN82

Specifications

  • No. of wells:
    96
  • Skirt:
    Semi-Skirted
  • Well volume:
    0.2 ml
  • Description:
    PCR plate, white, 10 per pack, 100 per case
  • Environmentally Preferable:
  • Cat. No.:
    104027-780

Specifications

About this item

Conduct efficient PCR assays with Alkali Scientific™ ProCycle™ 0.2 ml 96 well PCR plates, featuring semi skirt design in white.

  • Versatile thermal cycler compatibility: Compatible with a wide range of thermal cyclers
  • Effortless well identification: Featuring high-contrast, black alphanumeric indexing for easy well recognition
  • Optimal heat transfer: The ultra-thin, uniform well design ensures maximum heat transfer, enhancing reaction efficiency
  • Minimized sample evaporation: Well ridges help reduce sample evaporation when using film, foil, or strip caps for sealing
  • Premium materials: Crafted from high-clarity, ultra-pure polypropylene, meeting FDA 21 CFR 177.1520 and USP Class VI standards
  • Ideal for various industries: Suitable for applications in the pharmaceutical, food and molecular biology industries
  • Quality assurance: Certified free of detectable DNase, RNase, DNA, PCR inhibitors and rigorously tested for pyrogen-free performance

Enhance your PCR experiments with ProCycle™ 0.2 ml 96 well PCR plates. Featuring a semi-skirt design and white color, these plates offer versatility and clarity for molecular biology workflows. The semi-skirted design provides stability and compatibility with various PCR instruments, ensuring reliable performance in your experiments. With the high-quality construction and convenient packaging, these PCR plates are essential tools for efficient PCR amplification and analysis.

Packaging: Each pack contains 10 plates, and each case includes 100 plates, providing ample supply for your laboratory needs.